Butyl 2,2,3,3-tetrafluoropropyl carbonate
Fornitore: Apollo Scientific
APOSPC450447-25GEA
1840
CHF
APOSPC450447-25G
Butyl 2,2,3,3-tetrafluoropropyl carbonate
Butyl 2,2,3,3-tetrafluoropropyl carbonate
Formula:
C₈H₁₂F₄O₃ Peso molecolare: 232,173 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28334421 CAS Number: 1980034-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...