Stampa…

Ethyl 3-fluoropropyl carbonate

Fornitore: Apollo Scientific

APOSPC450409-25GEA 1840 CHF
APOSPC450409-25G
Ethyl 3-fluoropropyl carbonate
Ethyl 3-fluoropropyl carbonate
Formula: C₆H₁₁FO₃
Peso molecolare: 150,148 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334386
CAS Number: 888974-56-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR