Stampa…

Methyl 1H,1H,2H,2H-perfluorohexyl carbonate

Fornitore: Apollo Scientific

APOSPC450403-25GEA 1840 CHF
APOSPC450403-25G
Methyl 1H,1H,2H,2H-perfluorohexyl carbonate
Methyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formula: C₈H₇F₉O₃
Peso molecolare: 322,125 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334381
CAS Number: 1980076-59-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR