Stampa…

2,2,3,3,4,4,4-Heptafluorobutyl methyl carbonate

Fornitore: Apollo Scientific

APOSPC450399-25GEA 1840 CHF
APOSPC450399-25G
2,2,3,3,4,4,4-Heptafluorobutyl methyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl methyl carbonate
Formula: C₆H₅F₇O₃
Peso molecolare: 258,091 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334377
CAS Number: 277332-95-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR