Stampa…

4-Bromo-3,3,4,4-tetrafluorobutyl 3-fluoropropyl carbonate

Fornitore: Apollo Scientific

APOSPC450388-25GEA 1840 CHF
APOSPC450388-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 3-fluoropropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 3-fluoropropyl carbonate
Formula: C₈H₁₀BrF₅O₃
Peso molecolare: 329,059 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334460
CAS Number: 1923065-43-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR