1H,1H,7H-Perfluoroheptyl 1H,1H,2H,2H-perfluorohexyl carbonate
Fornitore: Apollo Scientific
APOSPC450376-25GEA
1840
CHF
APOSPC450376-25G
1H,1H,7H-Perfluoroheptyl 1H,1H,2H,2H-perfluorohexyl carbonate
1H,1H,7H-Perfluoroheptyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formula:
C₁₄H₇F₂₁O₃ Peso molecolare: 622,17 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28334369 CAS Number: 1980085-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...