Stampa…

1H,1H,9H-Perfluorononyl hexafluoroisopropyl carbonate

Fornitore: Apollo Scientific

APOSPC450366-25GEA 1840 CHF
APOSPC450366-25G
1H,1H,9H-Perfluorononyl hexafluoroisopropyl carbonate
1H,1H,9H-Perfluorononyl hexafluoroisopropyl carbonate
Formula: C₁₃H₄F₂₂O₃
Peso molecolare: 626,134 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334359
CAS Number: 1980048-97-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR