Stampa…

2,2-Difluoroethyl 1H,1H,2H,2H-perfluorohexyl carbonate

Fornitore: Apollo Scientific

APOSPC450333-25GEA 1840 CHF
APOSPC450333-25G
2,2-Difluoroethyl 1H,1H,2H,2H-perfluorohexyl carbonate
2,2-Difluoroethyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formula: C₉H₇F₁₁O₃
Peso molecolare: 372,133 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334326
CAS Number: 1980048-64-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR