Stampa…

2,2,2-Trifluoroethyl 1H,1H-perfluorohexyl carbonate

Fornitore: Apollo Scientific

APOSPC450328-25GEA 1840 CHF
APOSPC450328-25G
2,2,2-Trifluoroethyl 1H,1H-perfluorohexyl carbonate
2,2,2-Trifluoroethyl 1H,1H-perfluorohexyl carbonate
Formula: C₉H₄F₁₄O₃
Peso molecolare: 426,104 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334321
CAS Number: 1980076-13-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR