2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
Fornitore: Apollo Scientific
APOSPC450321-25GEA
1840
CHF
APOSPC450321-25G
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
Formula:
C₈H₅F₁₁O₃ Peso molecolare: 358,106 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28334314 CAS Number: 1980048-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...