2,2-Difluoroethyl 3-fluoropropyl carbonate
Fornitore: Apollo Scientific
APOSPC450302-25GEA
1840
CHF
APOSPC450302-25G
2,2-Difluoroethyl 3-fluoropropyl carbonate
2,2-Difluoroethyl 3-fluoropropyl carbonate
Formula:
C₆H₉F₃O₃ Peso molecolare: 186,129 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28334296 CAS Number: 1980044-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...