2,2-Difluoroethyl 2,2,3,3,3-pentafluoropropyl carbonate
Fornitore: Apollo Scientific
APOSPC450298-25GEA
1840
CHF
APOSPC450298-25G
2,2-Difluoroethyl 2,2,3,3,3-pentafluoropropyl carbonate
2,2-Difluoroethyl 2,2,3,3,3-pentafluoropropyl carbonate
Formula:
C₆H₅F₇O₃ Peso molecolare: 258,091 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28334292 CAS Number: 1980076-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...