Stampa…

5-Bromo-4-chloro-4,5,5-trifluoropentanamide

Fornitore: Apollo Scientific

APOSPC450092-5GEA 516 CHF
APOSPC450092-5G
5-Bromo-4-chloro-4,5,5-trifluoropentanamide
5-Bromo-4-chloro-4,5,5-trifluoropentanamide
Formula: C₅H₆BrClF₃NO
Peso molecolare: 268,459 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18074380
CAS Number: 155630-24-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR