1,1,1-Trifluoro-3-methylpentane
Fornitore: Apollo Scientific
APOSPC450075-5GEA
0
CHF
APOSPC450075-5G
1,1,1-Trifluoro-3-methylpentane
1,1,1-Trifluoro-3-methylpentane
Formula:
C₆H₁₁F₃ Peso molecolare: 140,147 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18074323 CAS Number: 20733-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...