Stampa…

1,1,1,2,2,6,6,7,7,7-Decafluoro-5-hydroxy-5-methylheptan-3-one

Fornitore: Apollo Scientific

APOSPC450042-1GEA 163 CHF
APOSPC450042-1G
1,1,1,2,2,6,6,7,7,7-Decafluoro-5-hydroxy-5-methylheptan-3-one
1,1,1,2,2,6,6,7,7,7-Decafluoro-5-hydroxy-5-methylheptan-3-one
Formula: C₈H₆F₁₀O₂
Peso molecolare: 324,116 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22123975
CAS Number: 6742-92-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR