Stampa…

(E)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile

Fornitore: Apollo Scientific

APOSPC450005-100MGEA 342 CHF
APOSPC450005-100MG
(E)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
(E)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
Formula: C₁₀H₈FN
Peso molecolare: 161,176 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22124012
CAS Number: 1365836-27-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR