3-Chloro-2-fluoro-4-methylbenzaldehyde 95+%
Fornitore: Apollo Scientific
APOSPC446196-1GEA
759
CHF
APOSPC446196-1G
3-Chloro-2-fluoro-4-methylbenzaldehyde 95+%
3-Chloro-2-fluoro-4-methylbenzaldehyde
Formula:
C₈H₆ClFO Peso molecolare: 172,584 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28751754 CAS Number: 1383951-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...