6-Fluoronicotinamide 95+%
Fornitore: Apollo Scientific
APOSPC446185-1GEA
621
CHF
APOSPC446185-1G
6-Fluoronicotinamide 95+%
6-Fluoronicotinamide
Formula:
C₆H₅FN₂O Peso molecolare: 140,115 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01755146 CAS Number: 369-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...