Stampa…

4-(2-Fluorophenyl)pyridine-2-carbonitrile 95+%

Fornitore: Apollo Scientific

APOSPC446154-1GEA 747 CHF
APOSPC446154-1G
4-(2-Fluorophenyl)pyridine-2-carbonitrile 95+%
4-(2-Fluorophenyl)pyridine-2-carbonitrile
Formula: C₁₂H₇FN₂
Peso molecolare: 198,196 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28337804
CAS Number: 1219454-74-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR