Stampa…

5-Fluoropyridine-2-carboxylic acid tert-butyl ester 95+%

Fornitore: Apollo Scientific

APOSPC446142-1GEA 621 CHF
APOSPC446142-1G
5-Fluoropyridine-2-carboxylic acid tert-butyl ester 95+%
5-Fluoropyridine-2-carboxylic acid tert-butyl ester
Formula: C₁₀H₁₂FNO₂
Peso molecolare: 197,206 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18910562
CAS Number: 1379037-04-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR