5-(4-Fluorophenyl)-2-methylfuran-3-carbonitrile 95+%
Fornitore: Apollo Scientific
APOSPC446126-1GEA
495
CHF
APOSPC446126-1G
5-(4-Fluorophenyl)-2-methylfuran-3-carbonitrile 95+%
5-(4-Fluorophenyl)-2-methylfuran-3-carbonitrile
Formula:
C₁₂H₈FNO Peso molecolare: 201,196 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD27937113 CAS Number: 1501382-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...