Stampa…

C-[2-Methyl-5-[4-(trifluoromethyl)phenyl]furan-3-yl]methylamine 95+%

Fornitore: Apollo Scientific

APOSPC446121-1GEA 621 CHF
APOSPC446121-1G
C-[2-Methyl-5-[4-(trifluoromethyl)phenyl]furan-3-yl]methylamine 95+%
C-[2-Methyl-5-[4-(trifluoromethyl)phenyl]furan-3-yl]methylamine
Formula: C₁₃H₁₂F₃NO
Peso molecolare: 255,236 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD27937108
CAS Number: 1858242-39-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR