Stampa…

[4-(4-Fluorophenyl)pyridin-2-yl]methanol 95+%

Fornitore: Apollo Scientific

APOSPC446112-1GEA 495 CHF
APOSPC446112-1G
[4-(4-Fluorophenyl)pyridin-2-yl]methanol 95+%
[4-(4-Fluorophenyl)pyridin-2-yl]methanol
Formula: C₁₂H₁₀FNO
Peso molecolare: 203,212 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16607075
CAS Number: 742682-91-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR