Stampa…

1-[4-(6-Dimethylamino-4-trifluoromethyl-pyridin-2-yl)-phenyl]-ethanone

Fornitore: Apollo Scientific

APOSPC446057-500MGEA 318 CHF
APOSPC446057-500MG
1-[4-(6-Dimethylamino-4-trifluoromethyl-pyridin-2-yl)-phenyl]-ethanone
1-[4-(6-Dimethylamino-4-trifluoromethyl-pyridin-2-yl)-phenyl]-ethanone
Formula: C₁₆H₁₅F₃N₂O
Peso molecolare: 308,298 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD19981356
CAS Number: 1311280-01-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR