Stampa…

3-(Difluoromethoxy)phenol 95%

Fornitore: Apollo Scientific
Pericolo

APOSPC4356-1GEA 126 CHF
APOSPC4356-1G
3-(Difluoromethoxy)phenol 95%
3-(Difluoromethoxy)phenol

Not to be stored in glass

Formula: C₇H₆F₂O₂
Peso molecolare: 160,118 g/mol
Flash Pt: >110 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00236235
CAS Number: 88798-13-4
UN: 2810
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR