(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone
Fornitore: Apollo Scientific
APOSPC430586-1GEA
691
CHF
APOSPC430586-1G
(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone
(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone
Formula:
C₁₄H₉ClFIO₂ Peso molecolare: 390,576 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11520937 CAS Number: 1233025-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...