Stampa…

N-Trifluoroacetyl-l-phenylalanine

Fornitore: Apollo Scientific
Attenzione

APOSPC430575-1GEA 628 CHF
APOSPC430575-1G
N-Trifluoroacetyl-l-phenylalanine
N-Trifluoroacetyl-l-phenylalanine
Formula: C₁₁H₁₀F₃NO₃
Peso molecolare: 261,197 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00800548
CAS Number: 350-09-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR