Stampa…

4-(Trifluoromethyl)-6-(furan-2-yl)pyrimidine-2-thiol

Fornitore: Apollo Scientific

APOSPC430533-100MGEA 303 CHF
APOSPC430533-100MG
4-(Trifluoromethyl)-6-(furan-2-yl)pyrimidine-2-thiol
4-(Trifluoromethyl)-6-(furan-2-yl)pyrimidine-2-thiol
Formula: C₉H₅F₃N₂OS
Peso molecolare: 246,209 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01929341
CAS Number: 329710-06-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR