Stampa…

Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate 95%

Fornitore: Apollo Scientific
Attenzione

APOSPC421234-1GEA 123 CHF
APOSPC421234-1G
Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate 95%
Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate
Formula: C₆H₉F₃O₂
Peso molecolare: 170,13 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18253569
CAS Number: 1188911-72-9
UN: 1993
ADR: 3,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR