Stampa…

2-Bromo-5-(trifluoromethyl)isonicotinic acid 97%

Fornitore: Apollo Scientific

APOSPC421159-250MGEA 220 CHF
APOSPC421159-250MG
2-Bromo-5-(trifluoromethyl)isonicotinic acid 97%
2-Bromo-5-(trifluoromethyl)isonicotinic acid
Formula: C₇H₃BrF₃NO₂
Peso molecolare: 270,003 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16610250
CAS Number: 823222-03-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR