Stampa…

5-Ethyl-2-fluorophenylboronic acid

Fornitore: Apollo Scientific
Attenzione

APOSPC412139-1GEA 332 CHF
APOSPC412139-1G
5-Ethyl-2-fluorophenylboronic acid
5-Ethyl-2-fluorophenylboronic acid
Formula: C₈H₁₀BFO₂
Peso molecolare: 167,973 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD18783131
CAS Number: 900175-03-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR