Stampa…

1-(4-Fluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine

Fornitore: Apollo Scientific

APOSPC410465-1GEA 994 CHF
APOSPC410465-1G
1-(4-Fluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
1-(4-Fluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
Formula: C₁₂H₁₄FN₃
Peso molecolare: 219,258 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04968191
CAS Number: 1006464-87-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR