Stampa…

3-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Fornitore: Apollo Scientific

APOSPC410417-1GEA 777 CHF
APOSPC410417-1G
3-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formula: C₇H₆BrF₃N₂O₂
Peso molecolare: 287,034 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04969244
CAS Number: 1006348-79-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR