Stampa…

3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine

Fornitore: Apollo Scientific

APOSPC410397-1GEA 620 CHF
APOSPC410397-1G
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine
Formula: C₁₀H₁₆F₃N₃
Peso molecolare: 235,249 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06805085
CAS Number: 1006340-62-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR