Stampa…

3-[1',3'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine

Fornitore: Apollo Scientific

APOSPC410378-1GEA 620 CHF
APOSPC410378-1G
3-[1',3'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
3-[1',3'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
Formula: C₁₂H₁₆F₃N₅
Peso molecolare: 287,284 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08556263
CAS Number: 1006323-05-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR