Stampa…

1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carbohydrazide

Fornitore: Apollo Scientific

APOSPC410314-1GEA 280 CHF
APOSPC410314-1G
1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carbohydrazide
1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carbohydrazide
Formula: C₁₂H₁₁F₃N₄O₂
Peso molecolare: 300,237 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04967948
CAS Number: 1004193-88-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR