Stampa…

1-(2-Chloro-6-fluorobenzyl)-4-nitro-1H-pyrazole

Fornitore: Apollo Scientific

APOSPC410308-1GEA 463 CHF
APOSPC410308-1G
1-(2-Chloro-6-fluorobenzyl)-4-nitro-1H-pyrazole
1-(2-Chloro-6-fluorobenzyl)-4-nitro-1H-pyrazole
Formula: C₁₀H₇ClFN₃O₂
Peso molecolare: 255,633 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02575856
CAS Number: 333444-49-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR