Stampa…

3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Fornitore: Apollo Scientific

APOSPC410284-1GEA 463 CHF
APOSPC410284-1G
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formula: C₁₀H₁₂ClF₃N₄O
Peso molecolare: 296,677 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03419755
CAS Number: 1001518-98-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR