Stampa…

2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Fornitore: Apollo Scientific

APOSPC410279-1GEA 280 CHF
APOSPC410279-1G
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formula: C₉H₁₁F₃N₄O
Peso molecolare: 248,205 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03419742
CAS Number: 1001518-93-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR