Stampa…

4-(Cyclohexyloxy)-3-fluoroaniline hydrochloride

Fornitore: Apollo Scientific

APOSPC410116-1GEA 620 CHF
APOSPC410116-1G
4-(Cyclohexyloxy)-3-fluoroaniline hydrochloride
4-(Cyclohexyloxy)-3-fluoroaniline hydrochloride
Formula: C₁₂H₁₆FNO·ClH
Peso molecolare: 245,721 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11499050
CAS Number: 1197234-19-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR