Stampa…

1-[3-(Trifluoromethyl)benzyl]-1H-pyrazol-4-amine dihydrochloride

Fornitore: Apollo Scientific

APOSPC410013-1GEA 463 CHF
APOSPC410013-1G
1-[3-(Trifluoromethyl)benzyl]-1H-pyrazol-4-amine dihydrochloride
1-[3-(Trifluoromethyl)benzyl]-1H-pyrazol-4-amine dihydrochloride
Formula: C₁₁H₁₀F₃N₃·2ClH
Peso molecolare: 314,134 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04967279
CAS Number: 1002033-51-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR