Stampa…

trans-4-Fluoro-L-Proline methyl ester hydrochloride 95+%

Fornitore: Apollo Scientific

APOSPC409019-250MGEA 371 CHF
APOSPC409019-250MG
trans-4-Fluoro-L-Proline methyl ester hydrochloride 95+%
trans-4-Fluoro-L-Proline methyl ester hydrochloride
Formula: C₆H₁₀FNO₂·ClH
Peso molecolare: 183,608 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07374354
CAS Number: 58281-80-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR