Stampa…

1-Nitro-3-(pentafluoroethyl)benzene

Fornitore: Apollo Scientific

APOSPC408831-1GEA 2430 CHF
APOSPC408831-1G
1-Nitro-3-(pentafluoroethyl)benzene
1-Nitro-3-(pentafluoroethyl)benzene
Formula: C₈H₄F₅NO₂
Peso molecolare: 241,115 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22053411
CAS Number: 656-84-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR