Stampa…

4-Chloro-5-fluoro-6-methylpyrimidine

Fornitore: Apollo Scientific

APOSPC408559-1GEA 545 CHF
APOSPC408559-1G
4-Chloro-5-fluoro-6-methylpyrimidine
4-Chloro-5-fluoro-6-methylpyrimidine
Formula: C₅H₄ClFN₂
Peso molecolare: 146,55 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16988365
CAS Number: 898044-55-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR