Stampa…

6-Fluoro-5-methyl-4-pyrimidinamine 95+%

Fornitore: Apollo Scientific

APOSPC408550-250MGEA 102 CHF
APOSPC408550-250MG
6-Fluoro-5-methyl-4-pyrimidinamine 95+%
6-Fluoro-5-methyl-4-pyrimidinamine
Formula: C₅H₆FN₃
Peso molecolare: 127,12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10000602
CAS Number: 18260-69-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR