Stampa…

(4-Fluoro-benzyl)-[1-(4-fluoro-benzyl)-1H-benzoimidazol-2-yl]-amine

Fornitore: Apollo Scientific

APOSPC408386-500MGEA 1040 CHF
APOSPC408386-500MG
(4-Fluoro-benzyl)-[1-(4-fluoro-benzyl)-1H-benzoimidazol-2-yl]-amine
(4-Fluoro-benzyl)-[1-(4-fluoro-benzyl)-1H-benzoimidazol-2-yl]-amine
Formula: C₂₁H₁₇F₂N₃
Peso molecolare: 349,377 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09697750
CAS Number: 899809-05-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR