Stampa…

4-(3-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride

Fornitore: Apollo Scientific

APOSPC408376-500MGEA 742 CHF
APOSPC408376-500MG
4-(3-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
4-(3-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Formula: C₁₂H₁₆FNS·ClH
Peso molecolare: 261,786 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD21098956
CAS Number: 1353963-00-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR