Stampa…

4-Fluoro-N-piperidin-3-ylmethyl-benzenesulphonamide hydrochloride

Fornitore: Apollo Scientific

APOSPC408367-500MGEA 742 CHF
APOSPC408367-500MG
4-Fluoro-N-piperidin-3-ylmethyl-benzenesulphonamide hydrochloride
4-Fluoro-N-piperidin-3-ylmethyl-benzenesulphonamide hydrochloride
Formula: C₁₂H₁₇FN₂O₂S·ClH
Peso molecolare: 308,8 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD21098935
CAS Number: 1353971-00-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR