Stampa…

5-Fluoro-2-(piperidin-4-ylmethoxy)-pyrimidine hydrochloride

Fornitore: Apollo Scientific

APOSPC408363-500MGEA 663 CHF
APOSPC408363-500MG
5-Fluoro-2-(piperidin-4-ylmethoxy)-pyrimidine hydrochloride
5-Fluoro-2-(piperidin-4-ylmethoxy)-pyrimidine hydrochloride
Formula: C₁₀H₁₄FN₃O·ClH
Peso molecolare: 247,697 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD21098930
CAS Number: 1353965-96-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR