4-(4-Fluoro-benzenesulphonyl)-piperidine hydrochloride
Fornitore: Apollo Scientific
APOSPC408343-500MGEA
593
CHF
APOSPC408343-500MG
4-(4-Fluoro-benzenesulphonyl)-piperidine hydrochloride
4-(4-Fluoro-benzenesulphonyl)-piperidine hydrochloride
Formula:
C₁₁H₁₄FNO₂S·ClH Peso molecolare: 279,759 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18837553 CAS Number: 105283-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...